MAT180695179 Matrix Scientific

2,4,5-Trichloro-N-(2-thienylmethyl)aniline

  • MDL number MFCD10687710
  • Cas Number : 1036574-05-6
  • Molecular Weight : 292.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/031661.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031661.html
Availability details for
Quantity
  • Availability
  • Add to Cart