MAT180695187 Matrix Scientific

N-(sec-Butyl)-N-ethylamine

  • MDL number MFCD08691715
  • Cas Number : 21035-44-9
  • Molecular Weight : 101.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032206.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032206.html
Availability details for
Quantity
  • Availability
  • Add to Cart