MAT180695199 Matrix Scientific

2,4-Dichloro-N-(2-methylbenzyl)aniline

  • MDL number MFCD10687636
  • Cas Number : 940367-72-6
  • Molecular Weight : 266.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/031562.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031562.html
Availability details for
Quantity
  • Availability
  • Add to Cart