MAT180695214 Matrix Scientific

N-(4-Methoxybenzyl)-3-methylaniline

  • MDL number MFCD02040666
  • Cas Number : 141606-35-1
  • Molecular Weight : 227.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/031676.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031676.html
Availability details for
Quantity
  • Availability
  • Add to Cart