MAT180695232 Matrix Scientific

N-Cyclopentyl-N-hexylamine

  • MDL number MFCD10687875
  • Cas Number : 20007-10-7
  • Molecular Weight : 169.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/031852.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031852.html
Availability details for
Quantity
  • Availability
  • Add to Cart