MAT180695237 Matrix Scientific

(2-Chlorophenyl)-N-(2-thienylmethyl)methanamine

  • MDL number MFCD04051939
  • Cas Number : 66200-70-2
  • Molecular Weight : 237.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/031856.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031856.html
Availability details for
Quantity
  • Availability
  • Add to Cart