MAT180695247 Matrix Scientific

N-Ethyl-N-(4-phenoxyphenyl)amine

  • MDL number MFCD03210684
  • Cas Number : 171349-99-8
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 213-214� Torr
Product Link http://www.matrixscientific.com/031452.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/031452.html
Availability details for
Quantity
  • Availability
  • Add to Cart