MAT180695258 Matrix Scientific

2-Formylpyridoimidazole

  • MDL number MFCD06739247
  • Cas Number : 118000-43-4
  • Molecular Weight : 146.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-114
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/032277.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032277.html
Availability details for
Quantity
  • Availability
  • Add to Cart