MAT180695268 Matrix Scientific

3,5-Bis(methylsulfonyl)benzylamine hydrochloride

  • MDL number MFCD10688274
  • Cas Number : 1171787-85-1
  • Molecular Weight : 299.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032289.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032289.html
Availability details for
Quantity
  • Availability
  • Add to Cart