MAT180937546 Matrix Scientific

1-(2-Amino-ethyl)-piperidin-4-ol dihydrochloride

  • MDL number MFCD10687123
  • Cas Number : 110484-18-9
  • Molecular Weight : 217.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021298.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021298.html
Availability details for
Quantity
  • Availability
  • Add to Cart