MAT180937558 Matrix Scientific

3-([1,1'-Biphenyl]-4-ylmethoxy)piperidinehydrochloride

  • MDL number MFCD11100494
  • Cas Number : 1184989-56-7
  • Molecular Weight : 303.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023164.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023164.html
Availability details for
Quantity
  • Availability
  • Add to Cart