MAT180937562 Matrix Scientific

3-Ethylphenyl 4-piperidinyl ether hydrochloride

  • MDL number MFCD11100499
  • Cas Number : 1338974-43-8
  • Molecular Weight : 241.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023293.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023293.html
Availability details for
Quantity
  • Availability
  • Add to Cart