MAT180937572 Matrix Scientific

4-[2-(2-Naphthyloxy)ethyl]piperidine hydrochloride

  • MDL number MFCD11100513
  • Cas Number : 1185125-50-1
  • Molecular Weight : 291.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023494.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023494.html
Availability details for
Quantity
  • Availability
  • Add to Cart