MAT180937574 Matrix Scientific

Methyl (2S,4S)-4-(4-methoxyphenoxy)-2-pyrrolidinecarboxylate hydrochloride

  • MDL number MFCD08690632
  • Cas Number : 1217819-16-3
  • Molecular Weight : 287.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023665.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023665.html
Availability details for
Quantity
  • Availability
  • Add to Cart