MAT181424978 Matrix Scientific

2-[4-(2-Amino-ethyl)-piperazin-1-yl]-ethylaminetetrahydrochloride

  • MDL number MFCD11100462
  • Cas Number : 852392-58-6
  • Molecular Weight : 318.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point <300
Country of Origin USA
Product Link http://www.matrixscientific.com/010011.html

Safety Information

MSDS http://www.matrixscientific.com/010011.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart