MAT181739867 Matrix Scientific

2-Dimethylaminomethyl-benzylamine dihydrobromide

  • MDL number MFCD11100469
  • Cas Number : 1185113-24-9
  • Molecular Weight : 326.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020809.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020809.html
Availability details for
Quantity
  • Availability
  • Add to Cart