MAT181739869 Matrix Scientific

3-(1-Pyrrolidinyl)-8-azabicyclo[3.2.1]octanedihydrochloride

  • MDL number MFCD11100476
  • Cas Number : 1185300-31-5
  • Molecular Weight : 253.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021801.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021801.html
Availability details for
Quantity
  • Availability
  • Add to Cart