MAT181739871 Matrix Scientific

3-Chloro-4-(2-ethoxyethoxy)aniline hydrochloride

  • MDL number MFCD11100479
  • Cas Number : 1185304-55-5
  • Molecular Weight : 252.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022339.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022339.html
Availability details for
Quantity
  • Availability
  • Add to Cart