MAT181739873 Matrix Scientific

4-(2-Ethoxyethoxy)-3-(trifluoromethyl)anilinehydrochloride

  • MDL number MFCD11100482
  • Cas Number : 1185304-61-3
  • Molecular Weight : 285.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022488.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022488.html
Availability details for
Quantity
  • Availability
  • Add to Cart