MAT181739874 Matrix Scientific

4-[2-(2-Methoxyethoxy)ethoxy]-3-(trifluoromethyl)-phenylamine hydrochloride

  • MDL number MFCD11100483
  • Cas Number : 1185297-16-8
  • Molecular Weight : 315.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022489.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022489.html
Availability details for
Quantity
  • Availability
  • Add to Cart