MAT181739879 Matrix Scientific

3-[(4-Bromo-2-fluorophenoxy)methyl]piperidinehydrochloride

  • MDL number MFCD11100489
  • Cas Number : 1185298-10-5
  • Molecular Weight : 324.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023042.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023042.html
Availability details for
Quantity
  • Availability
  • Add to Cart