MAT182086454 Matrix Scientific

5-(Trifluoromethyl)-2-(4-trifluoromethylphenyl)-isoxazole-4-carboxylic acid

  • MDL number MFCD11100463
  • Cas Number : 886497-47-8
  • Molecular Weight : 327.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-185�c)
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/012581.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012581.html
Availability details for
Quantity
  • Availability
  • Add to Cart