MAT182086468 Matrix Scientific

4-Bromo-2-methylbenzotrifluoride

  • MDL number MFCD11100526
  • Molecular Weight : 239.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Boiling point 26-28�7mmHg
Product Link http://www.matrixscientific.com/032375.html

Safety Information

hcodes Flammable
MSDS http://www.matrixscientific.com/032375.html
Availability details for
Quantity
  • Availability
  • Add to Cart