MAT182086490 Matrix Scientific

5-Bromo-2,4-bis(trifluoromethyl)pyridine

  • MDL number MFCD11100546
  • Cas Number : 1099598-05-6
  • Molecular Weight : 294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/032409.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032409.html
Availability details for
Quantity
  • Availability
  • Add to Cart