MAT182086516 Matrix Scientific

4-(1H-Benzoimidazol-2-yl)-butylaminedihydrochloride

  • MDL number MFCD09832498
  • Cas Number : 1177300-09-2
  • Molecular Weight : 262.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/012710.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/012710.html
Availability details for
Quantity
  • Availability
  • Add to Cart