MAT182086535 Matrix Scientific

2-Chloro-4,5-bis-(trifluoromethyl)pyridine

  • MDL number MFCD00082556
  • Cas Number : 109919-25-7
  • Molecular Weight : 249.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/032376.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032376.html
Availability details for
Quantity
  • Availability
  • Add to Cart