MAT182086536 Matrix Scientific

1-Bromo-2-(2,2,2-trifluoroethyl)benzene

  • MDL number MFCD09996898
  • Cas Number : 1027513-81-0
  • Molecular Weight : 239.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/032377.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032377.html
Availability details for
Quantity
  • Availability
  • Add to Cart