MAT182086545 Matrix Scientific

2,2-Dimethy-3-hexanone

  • MDL number MFCD00027140
  • Cas Number : 5405-79-8
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/032354.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032354.html
Availability details for
Quantity
  • Availability
  • Add to Cart