MAT182086552 Matrix Scientific

Ethyl 2-fluoroisobutyrate

  • MDL number MFCD08062432
  • Cas Number : 55816-69-8
  • Molecular Weight : 134.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 120-126
Product Link http://www.matrixscientific.com/032433.html

Safety Information

hcodes Flammable/Toxic/Refrigerate/Lachrymator
MSDS http://www.matrixscientific.com/032433.html
Availability details for
Quantity
  • Availability
  • Add to Cart