MAT184121674 Matrix Scientific

(cis-racemic)-1-Benzylpyrrolidine-3,4-dicarboxylic acid monomethyl ester

  • MDL number MFCD09965876
  • Cas Number : 474317-64-1
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/018166.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/018166.html
Availability details for
Quantity
  • Availability
  • Add to Cart