MAT184121677 Matrix Scientific

4-(3,5-Dimethyl-1H-pyrazol-4-ylmethyl)-benzoicacid hydrochloride

  • MDL number MFCD11100940
  • Cas Number : 1240134-87-5
  • Molecular Weight : 266.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020445.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020445.html
Availability details for
Quantity
  • Availability
  • Add to Cart