MAT184121679 Matrix Scientific

C-(2-Methyl-2H-pyrazol-3-yl)-methylaminedihydrochloride

  • MDL number MFCD11506366
  • Cas Number : 1185299-87-9
  • Molecular Weight : 184.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021008.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021008.html
Availability details for
Quantity
  • Availability
  • Add to Cart