MAT184121684 Matrix Scientific

N,N-Diethyl-8-azabicyclo[3.2.1]octan-3-aminedihydrochloride

  • MDL number MFCD11506371
  • Cas Number : 1185301-72-7
  • Molecular Weight : 255.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021797.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021797.html
Availability details for
Quantity
  • Availability
  • Add to Cart