MAT184121685 Matrix Scientific

N-[3-(2-Aminophenoxy)phenyl]acetamidehydrochloride

  • MDL number MFCD11506372
  • Cas Number : 1185304-44-2
  • Molecular Weight : 278.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021836.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021836.html
Availability details for
Quantity
  • Availability
  • Add to Cart