MAT184121687 Matrix Scientific

3-Chloro-4-[2-(1-piperidinyl)ethoxy]phenylaminedihydrochloride

  • MDL number MFCD11506374
  • Cas Number : 1185297-48-6
  • Molecular Weight : 327.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022355.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022355.html
Availability details for
Quantity
  • Availability
  • Add to Cart