MAT184121688 Matrix Scientific

3-Chloro-4-(3-fluorophenoxy)aniline hydrochloride

  • MDL number MFCD11506375
  • Cas Number : 1185297-43-1
  • Molecular Weight : 274.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022375.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022375.html
Availability details for
Quantity
  • Availability
  • Add to Cart