MAT184121694 Matrix Scientific

N-[2-(4-Amino-3-methylphenoxy)ethyl]-N,N-diethylamine dihydrochloride

  • MDL number MFCD11506381
  • Cas Number : 1185176-52-6
  • Molecular Weight : 295.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022802.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022802.html
Availability details for
Quantity
  • Availability
  • Add to Cart