MAT184121699 Matrix Scientific

3-{[(3-Chlorobenzyl)oxy]methyl}piperidinehydrochloride

  • MDL number MFCD11506386
  • Cas Number : 1185296-31-4
  • Molecular Weight : 276.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/022972.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/022972.html
Availability details for
Quantity
  • Availability
  • Add to Cart