MAT184121702 Matrix Scientific

3-(Allyloxy)piperidine hydrochloride

  • MDL number MFCD11506389
  • Cas Number : 1185297-03-3
  • Molecular Weight : 177.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023151.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023151.html
Availability details for
Quantity
  • Availability
  • Add to Cart