MAT184121709 Matrix Scientific

4-[(4-Methylbenzyl)oxy]piperidine hydrochloride

  • MDL number MFCD11506396
  • Cas Number : 1185165-54-1
  • Molecular Weight : 241.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023263.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023263.html
Availability details for
Quantity
  • Availability
  • Add to Cart