MAT184121710 Matrix Scientific

4-(3-Isopropylphenoxy)piperidine hydrochloride

  • MDL number MFCD11506397
  • Cas Number : 1185298-29-6
  • Molecular Weight : 255.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023290.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023290.html
Availability details for
Quantity
  • Availability
  • Add to Cart