MAT184121749 Matrix Scientific

2,3-Dimethylbenzotrifluoride

  • MDL number MFCD11226579
  • Cas Number : 80245-28-9
  • Molecular Weight : 174.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Boiling point 68-69� Torr
Product Link http://www.matrixscientific.com/032578.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032578.html
Availability details for
Quantity
  • Availability
  • Add to Cart