MAT184121762 Matrix Scientific

3-(6-Methoxy-1H-benzoimidazol-2-yl)-propionic acid hydrochloride

  • MDL number MFCD11506444
  • Cas Number : 1185299-60-8
  • Molecular Weight : 256.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033064.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033064.html
Availability details for
Quantity
  • Availability
  • Add to Cart