MAT184121769 Matrix Scientific

5-(2-Chloro-phenyl)-2H-pyrazol-3-ylaminehydrochloride

  • MDL number MFCD11506469
  • Cas Number : 1031791-02-2
  • Molecular Weight : 230.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033203.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033203.html
Availability details for
Quantity
  • Availability
  • Add to Cart