MAT184121775 Matrix Scientific

Methyl-[2-(2-methyl-imidazol-1-yl)-ethyl]-aminedihydrate

  • MDL number MFCD11506481
  • Cas Number : 1185299-41-5
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/033271.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/033271.html
Availability details for
Quantity
  • Availability
  • Add to Cart