MAT184121792 Matrix Scientific

N-Hydroxyisobutyramidine

  • MDL number MFCD00464225
  • Cas Number : 35613-84-4
  • Molecular Weight : 102.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-64
Purity 95
Storage temp. Refrigerate
Country of Origin USA
Product Link http://www.matrixscientific.com/032531.html

Safety Information

hcodes Irritant/Refrigerate
MSDS http://www.matrixscientific.com/032531.html
Availability details for
Quantity
  • Availability
  • Add to Cart