MAT184121824 Matrix Scientific

3-(Isopentyloxy)aniline hydrochloride

  • MDL number MFCD11506361
  • Cas Number : 1135289-27-8
  • Molecular Weight : 215.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/017116.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/017116.html
Availability details for
Quantity
  • Availability
  • Add to Cart