MAT184121867 Matrix Scientific

2-(5-Amino-2-methyl-benzoimidazol-1-yl)-ethanoldihydrochloride

  • MDL number MFCD01596248
  • Cas Number : 1158771-09-5
  • Molecular Weight : 264.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032609.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032609.html
Availability details for
Quantity
  • Availability
  • Add to Cart