MAT184121869 Matrix Scientific

1-[2-(1H-Indol-3-yl)-ethyl]-5-oxo-pyrrolidine-3-carboxylic acid

  • MDL number MFCD00424339
  • Cas Number : 295344-74-0
  • Molecular Weight : 272.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032611.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032611.html
Availability details for
Quantity
  • Availability
  • Add to Cart