MAT184121878 Matrix Scientific

2,2,5,5-Tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD00568208
  • Cas Number : 76194-00-8
  • Molecular Weight : 169.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/032620.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/032620.html
Availability details for
Quantity
  • Availability
  • Add to Cart